Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Antibody (4F12), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N104328APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N104328APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N104328APC7 Supplier Novus Biologicals Supplier No. NBP104328APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PD-1 Monoclonal antibody specifically detects PD-1 in Human, Canine samples. It is validated for Flow Cytometry, Product Image.

Specifications

Antigen PD-1
Applications Flow Cytometry
Classification Monoclonal
Clone 4F12
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Gene Symbols PDCD1
Host Species Mouse
Immunogen Recombinant human PDCD1 (21-167aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5133.0
Test Specificity Anti-human PDCD1 mAb, clone 4F12, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PDCD1 protein.
Target Species Human, Canine
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.