Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Antibody (F12A8) - VHH - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB176515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176515 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176515 Supplier Novus Biologicals Supplier No. NBP312827
Add to Cart
Add to Cart
This item is not returnable. View return policy

Llama Monoclonal Antibody

PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen PD-1
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone F12A8
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS
Gene Alias CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Host Species Llama
Immunogen PD-1 antibody was raised against a recombinant protein corresponding to amino acids 25 - 167 of human PD-1.
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.