Learn More
Description
Specifications
Specifications
| Antigen | PD-1 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
| Classification | Monoclonal |
| Clone | J116 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry |
| Gene Alias | CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Gene Symbols | PDCD1 |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody was PD1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
