Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Antibody (J43), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358900 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358900 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358900 Supplier Novus Biologicals Supplier No. NBP143110MFV610
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)

Specifications

Antigen PD-1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone J43
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was PD1.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.