Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1/CD279 Mouse anti-Human, Mouse, Rat, Clone: 4H4D1, Proteintech
SDP

Mouse Monoclonal Antibody

$518.60

Specifications

Antigen PD-1/CD279
Clone 4H4D1
Concentration 1.21 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-100-3189
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
662201IG150UL
150 μL
Each of 1 for $518.60
Only null left
Add to Cart
 
Description

Description

This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases.
Specifications

Specifications

PD-1/CD279
1.21 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CD279, hPD 1, hPD l, PD1, PD-1, PD-1/CD279, PDCD1, programmed cell death 1, Protein PD 1, SLEB2
PDCD1
IgG2b
Protein A
Antibody
4H4D1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
Q02242, Q15116
18566, 301626, 5133
PD-1/CD279 Fusion Protein Ag12470
Primary
-20°C
PDCD1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.