Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Mouse anti-Human, Clone: EH33.1F9.G4.5, MilliporeSigma™
Mouse Monoclonal Antibody
$208.00 - $488.00
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | EH33.1F9.G4.5 |
| Concentration | Please refer to lot specific datasheet. |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABC1122-25
|
MilliporeSigma
MABC112225UG |
25 μg |
Each for $208.00
|
|
|||||
|
MABC1122100
|
MilliporeSigma
MABC1122100UG |
100 μg |
Each for $488.00
|
|
|||||
Description
Anti-PD-1, clone EH33 is a mouse monoclonal antibody that detects Programmed cell death protein 1 (PD-1) in human cells and has been tested for use in Immunohistochemistry (Paraffin).Specifications
| PD-1 | |
| Please refer to lot specific datasheet. | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| PDCD1,PD1 | |
| IgG2a κ | |
| Protein G Purified | |
| Clone EH33 specifically detects PD-1 in human cells. It targets an epitope with the extracellular domain. |
| EH33.1F9.G4.5 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Inflammation and Immunology | |
| NP_005009.2 | |
| Recombinant protein fragment corresponding to the extracellular domain of human PD-1 (as an Ig fusion protein). | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title