Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PD 102807
GREENER_CHOICESDP

Catalog No. 167110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-711-0 10 mg
16-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-711-0 Supplier R&D Systems™ Supplier No. 1671/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective M4 antagonist

Selective M4 muscarinic receptor antagonist. IC50 values are 91, 6559, 3441, 950 and 7412 nM for human M4, M1, M2, M3, and M5 receptors respectively.

Chemical Identifiers

CAS 23062-91-1
Molecular Formula C23H24N2O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 392.45
Purity >98%
Molecular Weight (g/mol) 392.455
Chemical Name or Material PD 102807
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.