Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PD 102807

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective M4 muscarinic receptor antagonist. IC50 values are 91, 6559, 3441, 950 and 7412 nM for human M4, M1, M2, M3, and M5 receptors respectively.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 392.45 |
Purity | >98% |
Molecular Weight (g/mol) | 392.455 |
Chemical Name or Material | PD 102807 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction