Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD 153035 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
An extremely potent inhibitor of epidermal growth factor (EGF) receptor tyrosine kinase, with an IC50 of 25 pM. Inhibits other purified tyrosine kinases only at micromolar or higher concentrations.
Specifications
Specifications
| Quantity | 50 mg |
| Synonym | pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride |
| Formula Weight | 396.67 |
| Percent Purity | >98% |
| Chemical Name or Material | PD 153035 hydrochloride |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction