Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PD 161570

Click to view available options
Quantity:
10 mg
50 mg
Description
PD 161570 is a selective FGFR inhibitor (IC50 values are 40, 262 and 3700 nM for FGFR, PDGFR and EGFR respectively). Inhibits FGFR receptor phosphorylation (IC50 = 622 nM) and growth of A121 cells in vitro.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | PD 161570 |
| Purity | 0.97 |
| Molecular Weight (g/mol) | 532.51 |
| Inhibitors | PD 161570 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction