Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 166285 dihydrochloride
GREENER_CHOICE

Catalog No. 37851
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-851 1 mg
37-851-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-851 Supplier Tocris Bioscience™ Supplier No. 3785/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent Src inhibitor; also inhibits FGFR1, PDGFRβ and Wee1

Potent inhibitor of the tyrosine kinases c-Src, fibroblast growth factor receptor 1 (FGFR1), and platelet-derived growth factor receptor β (PDGFRβ) (IC50 values are 8.4, 39.3 and 98.3 nM respectively). Also inhibits the checkpoint kinases Wee1 and Myt1; abolishes Cdc2 phosphorylation in numerous tumor cell lines and abrogates the G2 checkpoint.

Chemical Identifiers

CAS 212391-63-4
Molecular Formula C26H29Cl4N5O2

Specifications

Quantity 1 mg
Formula Weight 585.35
Purity >99%
Molecular Weight (g/mol) 585.351
Chemical Name or Material PD 166285 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.