Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 166793
GREENER_CHOICE

Catalog No. 252010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-201-0 10 mg
25-205-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-201-0 Supplier Tocris Bioscience™ Supplier No. 2520/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Broad spectrum MMP inhibitor

Broad spectrum MMP inhibitor. Displays high affinity for MMP-2, -3 and -13 (IC50 values are 4, 7 and 8 nM respectively) and exhibits > 750-fold selectivity over MMP-1, -7 and -9. Attenuates left ventricular remodelling and dysfunction in rat model of heart failure.

Chemical Identifiers

CAS 199850-67-4
Molecular Formula C17H18BrNO4S

Specifications

Quantity 10 mg
Formula Weight 412.3
Purity >99%
Molecular Weight (g/mol) 412.298
Chemical Name or Material PD 166793
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.