Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 168568 dihydrochloride
GREENER_CHOICESDP

Catalog No. 352910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-291-0 10 mg
35-295-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-291-0 Supplier Tocris Bioscience™ Supplier No. 3529/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective D4 antagonist

Potent and selective dopamine D4 receptor antagonist (Ki values are 8.8 and 1842 nM at D4 and D2 receptors respectively). Reverses amphetamine-stimulated locomotion in vivo and is orally active.

Chemical Identifiers

CAS 210688-56-5
Molecular Formula C22H29Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 422.39
Purity >98%
Molecular Weight (g/mol) 422.394
Chemical Name or Material PD 168568 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.