Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 98059
GREENER_CHOICE

Catalog No. p-5545957
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MEK inhibitor

PD 98059 is an inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal.

Specifications

CAS 167869-21-8
Quantity 1 mg
Molecular Formula 2-(2-Amino-3-methoxyphenyl)-4 H-1-benzopyran-4-one
Product Type PD 98059
Purity 98%
Molecular Weight (g/mol) M.W. (Observed): 267.28
Inhibitors PD 98059

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.