Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PD 98059

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
PD 98059 is an inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal.
Specifications
Specifications
| CAS | 167869-21-8 |
| Quantity | 10 mg |
| Molecular Formula | 2-(2-Amino-3-methoxyphenyl)-4 H-1-benzopyran-4-one |
| Product Type | PD 98059 |
| Purity | 98% |
| Molecular Weight (g/mol) | M.W. (Observed): 267.28 |
| Inhibitors | PD 98059 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction