Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-ECGF/Thymidine Phosphorylase Antibody (OTI7A6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N273300A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273300A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273300A647 Supplier Novus Biologicals Supplier No. NBP273300AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PD-ECGF/Thymidine Phosphorylase
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7A6
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase
Gene Symbols TYMP
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 311-482 of human PD-ECGF/Thymidine Phosphorylase (NP_001944) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 1890.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.