Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-ECGF/Thymidine Phosphorylase Antibody (rTYMP/3444), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB392405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392405 Supplier Novus Biologicals Supplier No. NBP308359V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PD-ECGF/Thymidine Phosphorylase Monoclonal antibody specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen PD-ECGF/Thymidine Phosphorylase
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rTYMP/3444
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase
Host Species Mouse
Immunogen Recombinant full-length human PD-ECGF/Thymidine Phosphorylase protein (Uniprot: P19971 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 1890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.