Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Antibody (130021R), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314389 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314389 Supplier Novus Biologicals Supplier No. FAB1561RPECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PD-L1 Monoclonal specifically detects PD-L1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PD-L1
Applications Flow Cytometry
Classification Monoclonal
Clone 130021R
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PD-L1/B7-H1, Phe19-Thr239, Accession # Q9NZQ7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 29126
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.