Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB004444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004444 Supplier Novus Biologicals Supplier No. NBP176769C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PD-L1 Polyclonal specifically detects PD-L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence, Knockdown Validated.

Specifications

Antigen PD-L1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence, KnockDown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Gene Symbols CD274
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the center of human PD-L1. The immunogen is located within amino acids 60-110 of PD-L1.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
Primary or Secondary Primary
Test Specificity PD-L1 antibody has no cross-reactivity to PD-L2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.