Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Antibody (MIH5), Biotin, Novus Biologicals™
SDP

Catalog No. NBP14350605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14350605 0.05 mg
NBP1435065 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14350605 Supplier Novus Biologicals Supplier No. NBP1435060.05MG

Rat Monoclonal Antibody has been used in 2 publications

PD-L1 Monoclonal specifically detects PD-L1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen PD-L1/B7-H1
Applications Flow Cytometry
Classification Monoclonal
Clone MIH5
Concentration 0.5 mg/mL
Conjugate Biotin
Dilution Flow Cytometry 0.25 ug/10^6 cells in 100 ul
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Gene Alias B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Gene Symbols CD274
Host Species Rat
Immunogen The immunogen for this antibody was B7H1.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29126
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.