Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Antibody (MIH5), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N143262PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N143262PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N143262PEC5 Supplier Novus Biologicals Supplier No. NBP143262PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PD-L1 Monoclonal specifically detects PD-L1 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen PD-L1
Applications Flow Cytometry
Classification Monoclonal
Clone MIH5
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Gene Symbols CD274
Host Species Rat
Immunogen The immunogen for this antibody was B7H1.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 29126.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.