Learn More
Description
Specifications
Specifications
| Antigen | PD-L1/B7-H1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MIH5 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry |
| Gene Alias | Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Gene Symbols | CD274 |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was B7H1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
