Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Antibody (PDL1/8222R), Novus Biologicals™
SDP

Catalog No. NB235101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB235101 100 μg
NB233984 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB235101 Supplier Novus Biologicals Supplier No. NBP320339100UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen PD-L1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PDL1/8222R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Host Species Rabbit
Immunogen Recombinant fragment (around aa190-290) of human PD-L1 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 29126
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.