Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 B7-H1, Armenian Hamster anti-Mouse, Clone: 10B5, Millipore Sigma™

Catalog No. MABC113125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC113125 25 μg
MABC1131100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC113125 Supplier MilliporeSigma Supplier No. MABC113125UG
Only null left
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Anti-PD-L1 B7-H1, clone 10B5, Cat. No. MABC1131, is an Armenian hamster monoclonal antibody that detects Programmed cell death 1 ligand 1 (PD-L1) and has been tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Inhibition Function assay.

Programmed cell death 1 ligand 1 (UniProt: Q9EP73; also known as PD-L1, PDCD1 ligand 1, Programmed death ligand 1, B7 homolog 1, B7-H1, CD274) is encoded by the Cd274 (also known as B7h1, Pdcd1l1, Pdcd1lg1, Pdl1) gene (Gene ID: 50533) in murine species. PD-L1 is a single-pass type I membrane protein that is highly expressed in the heart, thymus, skeletal muscle, and lung. Its expression is also reported on activated dendritic cells, B-cells, and macrophages. Numerous tumor cell lines of lymphoid origin also express PD-L1. PD-L1 expression can be up-regulated by interferon-gamma treatment in monocytes, B cells, and macrophages. PD-1 and its ligands, PD-L1 and PD-L2 play a key role in the maintenance of peripheral tolerance, a process by which the quiescence of autoreactive mature T cells is maintained. However, tumors and pathogens that cause chronic infections can exploit this pathway to escape T-cell mediated tumor-specific and pathogen-specific immunity. The effector functions of T-cells expressing PD-1 can be downregulated by PD-L1 or PD-L2 expressed by the tumor cells. PD-L1 is initially produced with signal peptide (aa. 1-18) sequence, the removal of which yields the mature protein with a large extracellular (aa. 19-239) domain, a transmembrane domain (aa 240-260), and a cytoplasmic domain (aa 261-290).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1 B7-H1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Inhibition Assays
Classification Monoclonal
Clone 10B5
Conjugate Unconjugated
Dilution Inhibits Activity/Function Analysis: A representative lot blocked the activity of PD-L1 B7-H1 and inhibited apoptosis of activated alloreactive 2C T cells. (Hirano, F., et. al. (2005). Cancer Res. 65(3):1089-96; Goldberg, M.V., et. al. (2007). Blood. 110(1):186-92; Joshi, T., et. al. (2009). PLoS Pathog. 5(5):e1000431; Liu, X., et. al. (2016). Sci Rep. 6:36722). Flow Cytometry Analysis: A representative lot detected PD-L1 B7-H1 in Flow Cytometry applications (Hirano, F., et. al. (2005). Cancer Res. 65(3):1089-96; Dong, H., et. al. (2004). Immunity. 20(3):327-36).
Formulation Purified armenian hamster monoclonal antibody IgG2-IgG3 in PBS without azide.
Gene Alias Programmed cell death 1 ligand 1;PDCD1 ligand 1;Programmed death ligand 1;B7 homolog 1;B7-H1;CD274
Gene Symbols Cd274;B7h1;Pdcd1l1;Pdcd1lg1;Pdl1
Host Species Armenian Hamster
Immunogen Recombinant fragment corresponding to the extracellular domain of Mouse Programmed cell death 1 ligand 1.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_068693.1
Test Specificity This Armenian hamster monoclonal antibody detects Programmed cell death 1 ligand 1 (PD-L1) in murine species.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.