Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PD-L1 Polyclonal Antibody

Catalog No. PIPA5122122
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122122 20 μL
PIPA5119355 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122122 Supplier Invitrogen™ Supplier No. PA5122122
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PD-L1 Polyclonal Antibody for Western Blot, ICC/IF

Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Cd274
Gene Accession No. Q9EP73, Q9NZQ7
Gene Alias A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
Gene Symbols Cd274
Host Species Rabbit
Immunogen Recombinant protein Programmed Cell Death Protein 1 Ligand 1. The antigen corresponds to amino acid range 24-234 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29126, 499342, 574058, 60533
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.