Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L2/B7-DC/PDCD1LG2 Antibody (TY25), HRP, Novus Biologicals™
SDP

Catalog No. NB10065332H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065332H 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065332H Supplier Novus Biologicals Supplier No. NB10065332H

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TY25
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Gene Symbols PDCD1LG2
Host Species Rat
Immunogen B7-DC transfected RAW264.7 cells.
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80380
Test Specificity NB100-65332 recognizes mouse CD273, a 42kD member of the B7 family, otherwise known as B7-dendritic cell (B7-DC), or programmed death ligand-2 (PD-L2). The expression of CD273 is restricted to a subpopulation of dendritic cells (DCs), and DCs and macrophages that have been stimulated with interferon gamma, GM-CSF or IL-4. CD273 has been identified as a ligand for PD-1(CD279), along with another B7 family member B7-H1 (CD274). Studies suggest that both CD273 and B7-H1 co-stimulate or inhibit T cell proliferation and cytokine production. Antibodies produced by clone TY25 are reported to block the binding of CD273 (B7-DC) to it's receptor.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.