Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L2/B7-DC/PDCD1LG2 Rabbit anti-Human, Clone: 18, Novus Biologicals™
SDP

Catalog No. NB020265 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020265 100 μg
NB020266 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020265 Supplier Novus Biologicals Supplier No. NBP289391100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 18
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:1000-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Host Species Rabbit
Immunogen Recombinant Human PD-L2/B7-DC/PDCD1LG2 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80380
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.