Learn More
PD2 (PAF1), Mouse anti-Human, Clone: F169-3B2, Millipore Sigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MABT1495100UG
 View more versions of this product
Catalog No. MABT1495100
Only null left
Add to Cart
Description
Description
Anti-PD2 (PAF1), clone F169-3B2, Cat. No. MABT1495, is mouse monoclonal antibody that detects Pancreatic differentiation protein 2 and has been tested for use in ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Pancreatic differentiation protein 2 (UniProt: Q8N7H5; also known as RNA polymerase II-associated factor 1 homolog, PD2, hPAF1) is encoded by the PAF1 (also known as PD2) gene (Gene ID: 54623) in human. PD2 is a nuclear protein that serves as a subunit of the RNA polymerase II associated factor (PAF) complex. It has multiple functions during transcription by RNA polymerase II. It has been implicated in regulation of development and maintenance of embryonic stem cell pluripotency. It associates with RNA polymerase II through interaction with POLR2A C-terminal domain non-phosphorylated and serine 2 and serine 5 phosphorylated forms. It is involved in transcriptional elongation, acting both independently and synergistically with TCEA1 and in cooperation with the DSIF complex and HTATSF1. Three isoforms of PD2 have been described that are produced by alternative splicing. PD2 has oncogenic property and is shown to be overexpressed in pancreatic cancer cells. In normal murine pancreas, its expression is restricted to acinar cells, however, its expression is increased in the ductal cells of mouse model of pancreatic cancer with advancing age. (Ref.: Dey, P., et al (2014). Oncotarget, 5(12); 4480-4491).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD2 (PAF1)
Monoclonal
Unconjugated
Purified mouse monoclonal antibody IgG1 in PBS without azide.
PAF1;PD2
KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal half of human Pancreatic differentiation protein 2 (PD2).
100 μg
Epigenetics & Nuclear Function
NP_061961
Human
Purified
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
F169-3B2
Immunocytochemistry Analysis: A representative lot detected PD2 (PAF1) in Immunocytochemistry applications (Dey, P., et. al. (2014). Oncotarget. 5(12):4480-91; Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).Immunoprecipitation Analysis: A representative lot immunoprecipitated PD2 (PAF1) in Immunoprecipitation applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).ELISA Analysis: A representative lot detected PD2 (PAF1) in ELISA applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).Immunohistochemistry Analysis: A representative lot detected PD2 (PAF1) in Immunohistochemistry applications (Dey, P., et. al. (2014). Oncotarget. 5(12):4480-91).Western Blotting Analysis: A representative lot detected PD2 (PAF1) in Western Blotting applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).
Pancreatic differentiation protein 2;RNA polymerase II-associated factor 1 homolog;hPAF1
Mouse
Protein G purified
RUO
Primary
Clone F169-3B2 detects Pancreatic differentiation protein 2 (PD2) in human. It targets an epitope with in 22 amino acids from the C-terminal half.
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.