Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD2 (PAF1), Mouse anti-Human, Clone: F169-3B2, Millipore Sigma™

Catalog No. p-7162983 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT149525 25 μg
MABT1495100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT149525 Supplier MilliporeSigma Supplier No. MABT149525UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PD2 (PAF1), clone F169-3B2, Cat. No. MABT1495, is mouse monoclonal antibody that detects Pancreatic differentiation protein 2 and has been tested for use in ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.

Pancreatic differentiation protein 2 (UniProt: Q8N7H5; also known as RNA polymerase II-associated factor 1 homolog, PD2, hPAF1) is encoded by the PAF1 (also known as PD2) gene (Gene ID: 54623) in human. PD2 is a nuclear protein that serves as a subunit of the RNA polymerase II associated factor (PAF) complex. It has multiple functions during transcription by RNA polymerase II. It has been implicated in regulation of development and maintenance of embryonic stem cell pluripotency. It associates with RNA polymerase II through interaction with POLR2A C-terminal domain non-phosphorylated and serine 2 and serine 5 phosphorylated forms. It is involved in transcriptional elongation, acting both independently and synergistically with TCEA1 and in cooperation with the DSIF complex and HTATSF1. Three isoforms of PD2 have been described that are produced by alternative splicing. PD2 has oncogenic property and is shown to be overexpressed in pancreatic cancer cells. In normal murine pancreas, its expression is restricted to acinar cells, however, its expression is increased in the ductal cells of mouse model of pancreatic cancer with advancing age. (Ref.: Dey, P., et al (2014). Oncotarget, 5(12); 4480-4491).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD2 (PAF1)
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F169-3B2
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A representative lot detected PD2 (PAF1) in Immunocytochemistry applications (Dey, P., et. al. (2014). Oncotarget. 5(12):4480-91; Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).Immunoprecipitation Analysis: A representative lot immunoprecipitated PD2 (PAF1) in Immunoprecipitation applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).ELISA Analysis: A representative lot detected PD2 (PAF1) in ELISA applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).Immunohistochemistry Analysis: A representative lot detected PD2 (PAF1) in Immunohistochemistry applications (Dey, P., et. al. (2014). Oncotarget. 5(12):4480-91).Western Blotting Analysis: A representative lot detected PD2 (PAF1) in Western Blotting applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias Pancreatic differentiation protein 2;RNA polymerase II-associated factor 1 homolog;hPAF1
Gene Symbols PAF1;PD2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal half of human Pancreatic differentiation protein 2 (PD2).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_061961
Test Specificity Clone F169-3B2 detects Pancreatic differentiation protein 2 (PD2) in human. It targets an epitope with in 22 amino acids from the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.