Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDAP1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB050549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050549 Supplier Novus Biologicals Supplier No. NBP297989AF750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

PDAP1 Polyclonal specifically detects PDAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDAP1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias HASPP28, PAP1PDGFA-associated protein 1, PAP28 kDa heat- and acid-stable phosphoprotein, PDGFA associated protein 1, PDGF-associated protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PDAP1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11333
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.