Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD4 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB056052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056052 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056052 Supplier Novus Biologicals Supplier No. NBP298857AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDCD4 Polyclonal specifically detects PDCD4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDCD4
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PDCD4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 27250
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.