Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD5 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176988 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176988 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176988 Supplier Novus Biologicals Supplier No. NBP176988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDCD5 Polyclonal specifically detects PDCD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDCD5
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. CAG33215
Gene Alias FLJ42784, MGC9294, programmed cell death 5, programmed cell death protein 5, Protein TFAR19, TF-1 cell apoptosis-related protein 19, TFAR19 novel apoptosis-related, TFAR19TF1 cell apoptosis-related gene 19
Gene Symbols PDCD5
Host Species Rabbit
Immunogen Antibody was raised against a 21 amino acid synthetic peptide from near the carboxy terminus of human PDCD5. The immunogen is located within the last 50 amino acids of PDCD5. Amino Acid Squence: KFNRRKVMDSDEDDDY
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9141
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.