Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD7 Antibody (46C713.2.1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10056540R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056540R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056540R Supplier Novus Biologicals Supplier No. NB10056540R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDCD7 Monoclonal specifically detects PDCD7 in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen PDCD7
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 46C713.2.1
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q8N8D1
Gene Alias apoptosis-related protein ES18, ES18programmed cell death protein 7, FLJ54213, hES18, MGC22015, programmed cell death 7, U11/U12 snRNP 59K
Gene Symbols PDCD7
Host Species Mouse
Immunogen The antibody was raised against a synthetic peptide of mouse PDCD-7 protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10081
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.