Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCL Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017173 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017173 Supplier Novus Biologicals Supplier No. NBP281862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDCL Polyclonal specifically detects PDCL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PDCL
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, ELISA
Formulation PBS
Gene Alias DKFZp564M1863, PhLP, phosducin-like, phosducin-like protein
Gene Symbols PDCL
Host Species Rabbit
Immunogen PDCL antibody was raised against a 16 amino acid peptide near the amino terminus of human PDCL. The immunogen is located within the first 50 amino acids of PDCL. Amino Acid Squence: SSEDEDSDHEDKDRGR
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5082
Test Specificity PDCL antibody is human, mouse and rat reactive. At least two isoforms of PDCL are known to exist. PDCL antibody is predicted to not cross-react with other members of the PDCL protein family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.