Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCL2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB164962 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164962 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164962 Supplier Novus Biologicals Supplier No. NBP281765F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDCL2 Polyclonal antibody specifically detects PDCL2 in Human, Mouse, Rat samples. It is validated for ELISA

Specifications

Antigen PDCL2
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias GCPHLP, phosducin-like 2, phosducin-like protein 2
Host Species Rabbit
Immunogen PDCL2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human PDCL2. The immunogen is located within the last 50 amino acids of PDCL2. Amino Acid Squence: NTSIHDDSDSSNSDNDT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 132954
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.