Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCL3 Antibody (8F9), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP242694UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242694UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242694UV Supplier Novus Biologicals Supplier No. NBP242694UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDCL3 Monoclonal specifically detects PDCL3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDCL3
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 8F9
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HTPHLP, IAP-associated factor VIAF1, MGC3062, PHLP3, phosducin-like 3, phosducin-like protein 3, PhPL3, VIAF, VIAF-1, VIAF1PHLP2A, Viral IAP-associated factor 1
Gene Symbols PDCL3
Host Species Mouse
Immunogen Recombinant human PDCL3 (1-239aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 79031
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.