Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE10A Antibody, Novus Biologicals™
SDP

Catalog No. NB300645005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300645005 0.05 mg
NB018162 0.025 mg
NB300645 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB300645005 Supplier Novus Biologicals Supplier No. NB3006450.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

PDE10A Polyclonal specifically detects PDE10A in Human, Monkey, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen PDE10A
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Dilution Western Blot > 1:500, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation > 1:250, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Tris/Glycine buffer, pH 7.4; cryoprotective agents; HEPES; stabilizing protein (BSA 0.5%), and glycerol (30%). with 0.02% sodium azide
Gene Accession No. Q9Y233
Gene Alias cAMP and cAMP-inhibited cGMP 3′-5′-cyclic phosphodiesterase 10A, dJ416F21.1 (phosphodiesterase 10A), EC 3.1.4, EC 3.1.4.17, EC 3.1.4.35, FLJ11894, FLJ25677, HSPDE10A, phosphodiesterase 10A, phosphodiesterase 10A1 (PDE10A1)
Gene Symbols PDE10A
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 300-350 on human PDE10A2 protein common to all PDE10A variants.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 10846
Test Specificity This has no cross-reactivity against any other PDE family members tested so far.
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.