Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE10A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$268.00 - $706.50
Specifications
Antigen | PDE10A |
---|---|
Concentration | 0.7 mg/mL |
Dilution | Western Blot > 1:500, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation > 1:250, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018162
![]() |
Novus Biologicals
NB3006450.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NB300645005
![]() |
Novus Biologicals
NB3006450.05MG |
0.05 mg |
Each for $426.00
|
|
|||||
NB300645
![]() |
Novus Biologicals
NB300645 |
0.1 mg |
Each for $706.50
|
|
|||||
Description
PDE10A Polyclonal specifically detects PDE10A in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
PDE10A | |
Western Blot > 1:500, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation > 1:250, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Rabbit | |
Q9Y233 | |
PDE10A | |
IgG | |
This has no cross-reactivity against any other PDE family members tested so far. |
0.7 mg/mL | |
Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
Unconjugated | |
Monkey, Mouse, Rat, Human | |
10846 | |
Synthetic peptide taken within amino acid region 300-350 on human PDE10A2 protein common to all PDE10A variants. | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title