Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE10A Antibody (OTI1C9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP273273A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273273A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273273A30 Supplier Novus Biologicals Supplier No. NBP273273AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE10A Monoclonal specifically detects PDE10A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PDE10A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C9
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cAMP and cAMP-inhibited cGMP 3′-5′-cyclic phosphodiesterase 10A, dJ416F21.1 (phosphodiesterase 10A), EC 3.1.4, EC 3.1.4.17, EC 3.1.4.35, FLJ11894, FLJ25677, HSPDE10A, phosphodiesterase 10A, phosphodiesterase 10A1 (PDE10A1)
Gene Symbols PDE10A
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human PDE10A expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10846.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.