Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE11A Antibody, Novus Biologicals™
SDP

Catalog No. NB10061669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061669 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061669 Supplier Novus Biologicals Supplier No. NB10061669
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PDE11A Polyclonal specifically detects PDE11A in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen PDE11A
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_058649.3
Gene Alias cAMP and cGMP cyclic nucleotide phosphodiesterase 11A, cAMP and cGMP phosphodiesterase 11A, cyclic nucleotide phosphodiesterase 11A, dual 3'-5'-cyclic-AMP and -GMP phosphodiesterase 11A, EC 3.1.4, EC 3.1.4.17, EC 3.1.4.35, FLJ23693, MGC133355, MGC133356, PDE11A1, PDE11A2, PDE11A3, phosphodiesterase 11A, PPNAD2
Gene Symbols PDE11A
Host Species Goat
Immunogen Peptide with sequence C-NSFKESMEKSSYSD corresponding to internal region according to NP_001070665.1, NP_001070826.1, NP_001070664.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50940
Test Specificity This antibody is expected to recognize all four reported isoforms (NP_058649.3 ; NP_001070665.1; NP_001070826.1; NP_001070664.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.