Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE1A Antibody (OTI7D5), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP273277R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273277R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273277R Supplier Novus Biologicals Supplier No. NBP273277R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE1A Monoclonal specifically detects PDE1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PDE1A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7D5
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 3′-5′ cyclic nucleotide phosphodiesterase, 61 kDa Cam-PDE, calcium/calmodulin-dependent 3′-5′-cyclic nucleotide phosphodiesterase 1A, calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, calmodulin-dependent phosphodiesterase, Cam-PDE 1A, EC 3.1.4, EC 3.1.4.17, hCam-1, HCAM1, HSPDE1A, MGC26303, phosphodiesterase 1A, calmodulin-dependent, phosphodiesterase-1A
Gene Symbols PDE1A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 259-545 of human PDE1A (NP_005010) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5136.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.