Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE1B Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB336138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336138 Supplier Novus Biologicals Supplier No. NBP297975MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE1B Polyclonal antibody specifically detects PDE1B in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen PDE1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 63 kDa Cam-PDE, calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1B, Cam-PDE 1B, EC 3.1.4, EC 3.1.4.17, PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase IB, calmodulin-dependent, Phosphodiesterase-1B, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PDE1B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5153
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.