Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE1B Antibody (OTI6E4), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N273278A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273278A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273278A647 Supplier Novus Biologicals Supplier No. NBP273278AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE1B Monoclonal specifically detects PDE1B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDE1B
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6E4
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 63 kDa Cam-PDE, calcium/calmodulin-dependent 3′-5′-cyclic nucleotide phosphodiesterase 1B, Cam-PDE 1B, EC 3.1.4, EC 3.1.4.17, PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase, PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1, phosphodiesterase 1B, calmodulin-dependent, phosphodiesterase IB, calmodulin-dependent, Phosphodiesterase-1B, presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B
Gene Symbols PDE1B
Host Species Mouse
Immunogen Full length recombinant protein of human PDE1B (NP_000915) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5153.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.