Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE2A Antibody (201), HRP, Novus Biologicals™
SDP

Catalog No. NB070459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070459 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070459 Supplier Novus Biologicals Supplier No. NBP290159H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PDE2A Monoclonal specifically detects PDE2A in Human samples. It is validated for ELISA.

Specifications

Antigen PDE2A
Applications ELISA
Classification Monoclonal
Clone 201
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias cGMP-dependent 3'-5'-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PDE2A (O00408-1; Glu215-His900).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5138
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.