Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE2A Antibody (OTI3A4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP273288A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273288A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273288A94 Supplier Novus Biologicals Supplier No. NBP273288AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE2A Monoclonal specifically detects PDE2A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDE2A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3A4
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cGMP-dependent 3′-5′-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated
Gene Symbols PDE2A
Host Species Mouse
Immunogen Full length recombinant protein of human PDE2A (NP_002590) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5138
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.