Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE3A Antibody, Novus Biologicals™
SDP

Catalog No. NB300673005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300673005 0.05 mg
NB018170 0.025 mg
NB300673 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB300673005 Supplier Novus Biologicals Supplier No. NB3006730.05MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PDE3A Polyclonal specifically detects PDE3A in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PDE3A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunoprecipitation 1:200
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Alias CGI-PDE, CGI-PDE A, cGMP-inhibited 3′-5′-cyclic phosphodiesterase A, cyclic GMP inhibited phosphodiesterase A, Cyclic GMP-inhibited phosphodiesterase A, EC 3.1.4, EC 3.1.4.17, phosphodiesterase 3A, cGMP-inhibited
Gene Symbols PDE3A
Host Species Goat
Immunogen Synthetic peptide (selective for only PDE3A variant).
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 5139
Test Specificity Does not cross reacts with PDE4B variants nor with any other PDEs.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.