Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE3A Antibody, Novus Biologicals™
SDP

Catalog No. NBP146182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146182 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146182 Supplier Novus Biologicals Supplier No. NBP146182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE3A Polyclonal specifically detects PDE3A in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PDE3A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias CGI-PDE, CGI-PDE A, cGMP-inhibited 3'-5'-cyclic phosphodiesterase A, cyclic GMP inhibited phosphodiesterase A, Cyclic GMP-inhibited phosphodiesterase A, EC 3.1.4, EC 3.1.4.17, phosphodiesterase 3A, cGMP-inhibited
Gene Symbols PDE3A
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 575 and 625 of human Phosphodiesterase 3A using the numbering given in entry NP_000912.3 (GeneID 5139).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5139
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.