Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4B Antibody (OTI1D12), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP273292V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273292V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273292V Supplier Novus Biologicals Supplier No. NBP273292V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE4B Monoclonal specifically detects PDE4B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDE4B
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1D12
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cAMP-specific (phosphodiesterase E4 dunce homolog, cAMP-specific 3′-5′-cyclic phosphodiesterase 4B, cAMP-specific phosphodiesterase-4 B isoform, DKFZp686F2182, Drosophila), dunce-like phosphodiesterase E4, EC 3.1.4, EC 3.1.4.17, MGC126529, PDE32, PDEIVB, phosphodiesterase 4B, cAMP-specific, phosphodiesterase 4B, cAMP-specific (dunce, phosphodiesterase 4B, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E4), phosphodiesterase E4 dunce homolog
Gene Symbols PDE4B
Host Species Mouse
Immunogen Full length recombinant protein of human PDE4B (NP_002591) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5142.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.