Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4B Monoclonal Antibody (OTI2B10), TrueMAB™, OriGene
SDP

Catalog No. 502540904
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0904 30 μL
50-167-4340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0904 Supplier OriGene Supplier No. TA503446S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE4B Monoclonal antibody specifically detects PDE4B in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PDE4B
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2B10
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Asns
Gene Accession No. P08243
Gene Alias 3',5'-cyclic AMP phosphodiesterase; cAMP-specific 3',5'-cyclic phosphodiesterase 4B; DKFZp686F2182; DPDE4; dunce; dunce-like phosphodiesterase E4; MGC126529; MGC93193; OTTHUMP00000046896; PDE32; PDE4/IVb; PDE4B; Pde4B1; Pde4B2; Pde4B3; Pde4B4; PDEIVB; phosphodiesterase 4B; Phosphodiesterase 4B cAMP-specific (dunce; phosphodiesterase 4B, cAMP specific; phosphodiesterase 4B, cAMP-specific; Phosphodiesterase 4B, cAMP-specific (dunce; phosphodiesterase 4B, cAMP-specific (phosphodiesterase E4 dunce homolog, Drosophila); R74983; RP5-876C12.1
Host Species Mouse
Immunogen Full length human recombinant protein of human ASNS produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.