Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4C Antibody, Novus Biologicals™
SDP

Catalog No. NBP219746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219746 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219746 Supplier Novus Biologicals Supplier No. NBP219746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE4C Polyclonal specifically detects PDE4C in Human samples. It is validated for Western Blot.

Specifications

Antigen PDE4C
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q08493
Gene Alias cAMP-specific 3'-5'-cyclic phosphodiesterase 4C, DPDE1dunce, Drosophila), EC 3.1.4, EC 3.1.4.17, MGC126222, PDE21, phosphodiesterase 4C, cAMP-specific, phosphodiesterase 4C, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E1), phosphodiesterase 4C, cAMP-specific (phosphodiesterase E1 dunce homolog, phosphodiesterase E1 dunce homolog
Gene Symbols PDE4C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PDE4C. The exact sequence is proprietary.
Molecular Weight of Antigen 80 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5143
Test Specificity Human. Species reactivity based on sequence identity: Mouse (90%), Bovine (95%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.